Learn More
TARGETMOL CHEMICALS INC PF-04979064 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T734325MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-04979064 is a potent and selective PI3K and mTOR dual kinase inhibitor(Ki of 0.13 nM and 1.42 nMrespectively). purity: 98%
Catalog No. 50-257-9208