Learn More
Medchemexpress LLC PF-04991532 | 1215197-37-7 | 99.4% | 396.36 | C18H19F3N4O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10018150MG
Encompass_Preferred
PF-04991532 is a hepatoselective glucokinase activator provided for research use. It exhibits potent activation of glucokinase (EC50 = 80 nM in human, 100 nM in rat) and has been evaluated in preclinical studies for glucose-lowering effects without causing hepatic steatosis. The compound is supplied as a high-purity solid with supporting documentation for research applications.
  • Potent hepatoselective glucokinase activator (EC50 80 nM human, 100 nM rat).
  • Characterized in preclinical studies for glucose-lowering without hepatic steatosis.
  • High purity solid suitable for in vitro and in vivo research.
  • Available in multiple pack sizes to support small- and large-scale studies.
  • Supplier provides data sheet, safety data sheet, and certificate of analysis.
Catalog No. 50-002-15699