Learn More
Medchemexpress LLC PF 05089771 | 1235403-62-9 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12883100MG
Encompass_Preferred
PF 05089771 is a potent, orally active, and selective arylsulfonamide Nav1.7 inhibitor. It is currently being studied for its potential use in pain management and diabetic neuropathy.
  • Potent, orally active, and selective arylsulfonamide Nav1.7 inhibitor
  • IC50 values of 11 nM for hNav1.7, 12 nM for cynNav1.7, 13 nM for dogNav1.7, 171 nM for ratNav1.7, and 8 nM for musNav1.7
  • More than 1000-fold selective over tetrodotoxin-resistant (TTX-R) Nav1.5 and Nav1.8 channels (IC50s >10 μM)
  • Exhibits a range of selectivity over TTX-sensitive (TTX-S) channels (10-fold for Nav1.2 to 900-fold for Nav1.3 and Nav1.4)
  • Blocks the majority of TTX-S current (75.5 ± 10.5%) at 30 nM, with complete block at 100 nM
Catalog No. 50-003-39659