Learn More
Apexbio Technology LLC PF-05212384 (PKI-587), 5mg. Cas: 1197160-78-3 MFCD: MFCD16875679. PI3Kα, PI3Kγ and mTOR inhibitor
SDP
Supplier:  Apexbio Technology LLC B21795
Encompass_Preferred
PF-05212384 is an inhibitor of PI3K/mTOR with IC50 values of 0.4nM, 6nM, 8nM, 6nM and 1.4nM for PI3K, PI3K, PI3K, PI3K and mTOR, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5324