Learn More
Medchemexpress LLC HY-113697 10mg Medchemexpress, PF-05231023 CAS:1037589-69-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11369710MG
Encompass_Preferred
Medchemexpress, HY-113697 10mg PF-05231023 CAS:1037589-69-7 PF-05231023, a long-acting fibroblast growth factor 21 (FGF21) analog, is a FGF21-receptor agonist, suitable for development as a potential treatment for T2DM[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9261