Learn More
Medchemexpress LLC PF-06305591 50mg | 1449473-97-5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11430150MG
Encompass_Preferred
PF-06305591 is a potent, highly selective blocker of the NaV1.8 voltage-gated sodium channel (IC50 = 15 nM). It is supplied as a high-purity solid suitable for preclinical in vitro pharmacology and ADME studies, with recommended cold storage to preserve stability.
  • Potent NaV1.8 blockade (IC50 = 15 nM)
  • High purity suitable for in vitro research
  • Solid powder form with recommended cold storage
  • Available in small-scale quantities for preclinical testing
  • Also available as a DMSO solution option for cell-based assays
Catalog No. 50-002-29424