Learn More
Selleck Chemical LLC PF-06409577 10mg 1467057-23-3
SDP
Supplier:  Selleck Chemical LLC S833510MG
Encompass_Preferred
PF-06409577 is orally bioavailable AMPK activator with EC50 of 7 nM for ?1?1?1 in the TR-FRET assay and it shows no detectable inhibition of hERG in a patch-clamp assay (100 ?M) and was not an inhibitor (IC50 > 100 ?M) of the microsomal activities of major human cytochrome P450 isoforms. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8276