Learn More
Medchemexpress LLC HY-108691 5mg Medchemexpress, PF-06465469 CAS:1407966-77-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086915MG
Encompass_Preferred
Medchemexpress, HY-108691 5mg PF-06465469 CAS:1407966-77-1 PF-06465469 is a covalent inhibitor of ITK with an IC50 of 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0751