Learn More
Medchemexpress LLC HY-19836 10mg Medchemexpress, PF-06650833 CAS:1817626-54-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1983610MG
Encompass_Preferred
Medchemexpress, HY-19836 10mg PF-06650833 CAS:1817626-54-2 PF-06650833 is a potent, selective and orally active inhibitor of interleukin-1 receptor associated kinase 4 (IRAK4) with IC50s of 0.2 and 2.4 nM in the cell and PBMC assay, respectively. PF-06650833 is used to treat diseases such as rheumatoid arthritis, lupus, and lymphomas[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9298