Learn More
Medchemexpress LLC PF-06761281 | 1854061-19-0 | 99.5% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12066950MG
Encompass_Preferred
Research-grade small-molecule inhibitor of the sodium-coupled citrate transporter (NaCT/SLC13A5) for in vitro and in vivo studies of citrate transport and metabolism. Supplied as an off-white to light yellow solid with high purity and characterized potency across cell-based and hepatocyte assays.
  • Potent NaCT inhibitor with cellular IC50 ≈ 0.51 μM.
  • Partial selectivity versus NaDC1 and NaDC3 (IC50 ≈ 13.2 μM and 14.1 μM).
  • Demonstrated activity in hepatocytes (rat 0.12 μM, mouse 0.21 μM, human 0.74 μM).
  • High purity solid, suitable for biochemical and pharmacology studies.
  • Soluble in aqueous media at experimental concentrations; store sealed at -20°C.
  • Available in small-mass quantities for research use.
Catalog No. 50-002-32741