Learn More
Medchemexpress LLC Pf-06795071 | 2075629-81-9 | 99.31% | 399.34 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111512
Encompass_Preferred
PF-06795071 is a potent and selective covalent MAGL inhibitor with an IC50 of 3 nM. It shows excellent selectivity against FAAH. This product is suitable for various laboratory applications including neuroinflammation research.
  • Potent and selective covalent MAGL inhibitor
  • IC50 of 3 nM against MAGL
  • Excellent selectivity against FAAH
  • White to off-white solid appearance
Catalog No. 50-003-46921