Learn More
Medchemexpress LLC PF-06815345 hydrochloride | 2334434-49-8 | 98.2% | 634.49 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112598A100MG
Encompass_Preferred
PF-06815345 hydrochloride is an orally active and potent inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) with an IC50 value of 13.4 μM. PF-06815345 hydrochloride significantly decreases the PCSK9 level in vivo in mouse.
  • Orally active and potent inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9).
  • IC50 value of 13.4 μM.
  • Significantly decreases the PCSK9 level in vivo in mouse.
Catalog No. 50-004-36320