Learn More
Cayman Chemical PF-06869206 5mg
SDP
Supplier:  Cayman Chemical 281585 MG
Encompass_Preferred
An NaPi2a inhibitor (IC50 = 380 nM); selective for NaPi2a over NaPi2b, NaPi2c, PiT-1, and PiT-2 (IC50s = >25 μM)
Catalog No. 50-294-0801