Learn More
Medchemexpress LLC Pf-07038124 hydrochloride | 2415317-57-4 | 99.26% | C18H23BClNO4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY144683A
Encompass_Preferred
PF-07038124 hydrochloride is a PDE4 inhibitor with an IC50 of 0.5 nM for PDE4B2. It shows inhibitory activities against IL-13, IL4, and IFNγ (IC50=125, 4.1, 1.06 nM, respectively). This compound can be used for research of atopic dermatitis and plaque psoriasis.
  • Inhibits PDE4 at 0.5 nM
  • Suitable for research of atopic dermatitis and plaque psoriasis
  • Has demonstrated inhibitory activities against IL-13, IL4, and IFNγ
  • For research use only
Catalog No. 50-003-56585