Learn More
Medchemexpress LLC PF-07038124 hydrochloride | 2415317-57-4 | 99.3% | C18H23BClNO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY144683A10MG
Encompass_Preferred
PF-07038124 hydrochloride is the hydrochloride salt of a potent phosphodiesterase-4 (PDE4) inhibitor developed for topical treatment research in atopic dermatitis and plaque psoriasis. Supplied as a high-purity off-white solid, it demonstrates nanomolar activity against PDE4B2 and modulates key cytokines in cellular assays.
  • High purity solid for research use.
  • Nanomolar potency against PDE4B2.
  • Modulates IL-13, IL-4, and IFNγ in cellular assays.
  • Suitable for topical formulation and preclinical studies.
  • Provided as a stable hydrochloride salt.
Catalog No. 50-002-59136