Learn More
Abcam PF 356231, MMP-12 inhibitor, 1MG
Supplier:  Abcam AB1452371MG
Encompass_Preferred
MW 428.5 Da, Purity >98%. Potent and selective non-chelating MMP-12 inhibitor (IC₅₀ = 14 nM). Inhibits other MMPs at higher concentrations. Inhibits cancer cell migration and invasion.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5371