Learn More
Selleck Chemical LLC PF-3716556 10mg 928774-43-0
SDP
Supplier:  Selleck Chemical LLC S222210MG
Encompass_Preferred
PF 3716556 is a potent and selective P-CAB (potassium-competitive acid blocker), with pIC50 of 6.026 and 7.095 for the inhibition of porcine H+,K+-ATPase activity in ion-leaky and ion-tight assay, respectively, inhibits gastric acid secretion, displays no activity at Na+,K+-ATPase, used for the treatment of gastroesophageal reflux disease. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-725-7