Learn More
Medchemexpress LLC PF-4191834 10mg | 1029317-21-2 | 393.50 g/mol | C22H23N3O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11704810MG
Encompass_Preferred
PF-4191834 is an orally active, selective 5-lipoxygenase (5-LOX) inhibitor provided for research use. It is supplied as a solid with high reported purity and is intended for in vitro and in vivo studies of inflammatory pathways and leukotriene biosynthesis.
  • Orally active 5-LOX inhibitor for inflammation research.
  • High reported purity suitable for analytical and biological assays.
  • Available in small milligram quantities and as ready-to-use DMSO solutions.
  • Solid, white to yellow appearance; soluble in DMSO.
  • Provided with analytical purity information for quality control.
Catalog No. 50-002-29524