Learn More
Selleck Chemical LLC PF 429242 947303-87-9
SDP
Supplier:  Selleck Chemical LLC S64185MG
Encompass_Preferred
PF 429242 is known as a S1P inhibitor with an IC50 of 170 nM, showing no significant inhibition of trypsin, elastase, proteinase K, plasmin, kallikren, factor XIa, thrombin, or furin at concentrations up to 100 ?M and only modest inhibition of urokinase (IC50 = 50 ?M) and factor Xa (IC50 = 100 ?M). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1838657