Learn More
Selleck Chemical LLC PF-431396 10mg 717906-29-1
SDP
Supplier:  Selleck Chemical LLC S764410MG
Encompass_Preferred
PF-431396 is a dual PYK2/FAK inhibitor with IC50 of 11 nM and 2 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5065