Learn More
TARGETMOL CHEMICALS INC PF-431396 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T231425MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-431396 is a dual PYK2/FAK inhibitor (IC50 11/2 nM). purity: 99%
Catalog No. 50-258-0895