Learn More
Apexbio Technology LLC PF-431396 717906-29-1 10mg
SDP
Supplier:  Apexbio Technology LLC A869210
Encompass_Preferred
PF-431396 is a pyrimidine-based small-molecule inhibitor targeting proline-rich tyrosine kinase 2 (Pyk2) and focal adhesion kinase (FAK) It is designed to selectively inhibit these kinases thereby modulating signaling pathways involved in cellular adhesion migration and proliferation PF-431396 exerts its biological activity primarily through disruption of Pyk2 and FAK tyrosine autophosphorylation or transphosphorylation Based on these pharmacological properties PF-431396 holds research potential in the investigation of cancer biology inflammation and immunological cell responses in experimental models
Catalog No. 50-000-95949