Learn More
Medchemexpress LLC PF-4618433 | 1166393-85-6 | 98.62% | 445.52 | 100 MG

Supplier: Medchemexpress LLC HY18312100MG
PF-4618433 is a potent and selective PYK2 inhibitor, with an IC50 of 637 nM. It may be suitable for the research of osteoporosis, craniofacial and appendicular skeletal defects, and for targeted bone regeneration.
- Potent and selective PYK2 inhibitor.
- IC50 of 637 nM against PYK2.
- Promotes osteogenesis of hMSC cultures.
- Increases alkaline phosphatase (ALP) activity and mineralization.
- Enhances osteoblast proliferation.
- Enhances calcium deposition.
- Molecular weight: 445.52.
- Formula: C24H27N7O2.
- Appearance: Solid, white to off-white.
- Storage: Powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month.
- Purity: 98.62%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.