Learn More
TARGETMOL CHEMICALS INC PF 477736 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T602810MG
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF 477736 (PF-736PF-00477736) is a specifc effective and ATP-competitive Chk1 inhibitor (Ki 0.49 nM ) and also inhibits FGFR3 Aurora-A VEGFR2 Flt3 Fms (CSF1R) Ret and Yes. purity: 98%
Catalog No. 50-257-6623