Learn More
TARGETMOL CHEMICALS INC PF-4981517 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2154810MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. PF-4981517 (CYP3cide) is a efficient specific and time-dependent inhibitor of cytochrome P4503A4 (CYP3A4). The IC 50 values for CYP3A activity are 0.03 uM17 uM and 71 uM for CYP3A4 CYP3A5 and CYP3A7 respectively. PF-4981517 is able to be used to distinguish the contributions of CYP3A4 versus CYP3A5 on drug Metabolism. purity: 99%
Catalog No. 50-258-1026