Learn More
Medchemexpress LLC PF-4989216 | 1276553-09-3 | 99.4% | 380.40 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1386450MG
Encompass_Preferred
PF-4989216 is a potent and selective PI3Kα inhibitor with a Ki of 0.6 nM. It is for research use only.
  • Potent and selective PI3Kα inhibitor (Ki of 0.6 nM)
  • Excellent selectivity against mTOR (Ki=1440 nM) and other kinases
  • Demonstrates dose-responsive tumor growth inhibitory activity in xenograft models
  • Available as white to off-white solid
  • Suitable for research use only
Catalog No. 50-003-51681