Learn More
Selleck Chemical LLC PF-562271 5mg 717907-75-0
SDP
Supplier:  Selleck Chemical LLC S28905MG
Encompass_Preferred
PF-562271 is a potent, ATP-competitive, reversible inhibitor of FAK with IC50 of 1.5 nM in cell-free assays, ~10-fold less potent for Pyk2 than FAK and >100-fold selectivity against other protein kinases, except for some CDKs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3139