Learn More
Medchemexpress LLC PF-562271 (hydrochloride) | 939791-41-0 | 98.10% | 543.95 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY204035MG
Encompass_Preferred
PF-562271 hydrochloride is a potent, ATP-competitive, and reversible inhibitor targeting FAK (Focal Adhesion Kinase) and Pyk2 (Proline-rich tyrosine kinase 2) kinases, with IC50 values of 1.5 nM and 13 nM, respectively.
  • Potent, ATP-competitive, and reversible kinase inhibitor.
  • Selectively targets FAK and Pyk2 kinases.
  • Exhibits IC50s of 1.5 nM for FAK and 13 nM for Pyk2.
  • Also inhibits CDK2/E, CDK5/p35, CDK1/B, and CDK3/E.
  • Impairs cell viability in various cell lines, including Ewing sarcoma.
  • Inhibits FAK phosphorylation in vivo in a dose-dependent manner.
  • Demonstrates positive effects on bone tumors, including decreased tumor growth and deposition of new bone.
Catalog No. 50-004-00564