Learn More
Apexbio Technology LLC PF-573228, 10mg. Cas: 869288-64-2 MFCD: MFCD11519967. ATP-competitive FAK inhibitor
SDP
Supplier:  Apexbio Technology LLC B152310
Encompass_Preferred
PF573228 is an inhibitor of FAK with IC50 value of 4 nM [1].FAK (Focal adhesion kinase) is a non-receptor protein-tyrosine kinase that resides at the sites of focal adhesions. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5251