Learn More
Selleck Chemical LLC PF-573228 S2013-50mg
SDP
Supplier:  Selleck Chemical LLC S201350MG
Encompass_Preferred
PF-573228 is an ATP-competitive inhibitor of FAK with IC50 of 4 nM in a cell-free assay 50- to 250-fold selective for FAK than Pyk2 CDK1/7 and GSK-3 PF-573228 induces apoptosis
Catalog No. 50-310-8116