Learn More
Medchemexpress LLC PF06650833 50MG
SDP
Supplier:  Medchemexpress LLC HY1983650MG
Encompass_Preferred
PF06650833 is a potent, selective and oral bioavailability inhibitor of interleukin-1 receptor associated kinase 4 (IRAK4) with an IC50 of 0.2 nM, and used to treat diseases such as rheumatoid arthritis, lupus, and lymphomas.Purity: 99.71%
Catalog No. NC1745690