Learn More
Medchemexpress LLC PF429242 dihydrochloride | 2248666-66-0 | 99.8% | C25H37Cl2N3O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13447A10MG
Encompass_Preferred
PF429242 dihydrochloride is a reversible, competitive inhibitor of SREBP site-1 protease (S1P) used in biochemical and cell-based research to block SREBP processing and probe lipid and cholesterol biosynthesis pathways.
  • Reversible, competitive S1P inhibitor (IC50 = 175 nM).
  • Suitable for biochemical and cell-based studies of SREBP processing and lipid regulation.
  • High reported purity: 99.8%.
  • Molecular formula C25H37Cl2N3O2; mol wt 482.49 g/mol.
  • Provided as the dihydrochloride salt for improved solubility.
Catalog No. 50-002-60142