Learn More
Medchemexpress LLC PF9601N | 133845-63-3 | 99.9% | 25 MG

Supplier: Medchemexpress LLC HY12041925MG
PF9601N is a selective monoamine oxidase B (MAO-B) inhibitor with reported neuroprotective activity in cellular and animal models of Parkinson's disease. The compound contains an alkyne functional group that enables copper-catalyzed azide-alkyne cycloaddition (click chemistry) and is supplied as a high-purity powder for research applications.
- Selective monoamine oxidase B inhibitor suitable for neurodegeneration research.
- Contains an alkyne moiety compatible with click chemistry reactions.
- High purity (99.85%) for reliable experimental results.
- Available in multiple milligram quantities for small-scale studies.
- Powder stable at -20°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.