Learn More
Medchemexpress LLC Pf9601n | 133845-63-3 | 99.9% | 50 MG

Supplier: Medchemexpress LLC HY12041950MG
PF9601N is a research-grade selective monoamine oxidase B (MAO-B) inhibitor with reported neuroprotective activity in preclinical models, supplied as an off-white to light yellow solid for laboratory research use only.
- Molecular formula: C19H18N2O.
- Molecular weight: 290.36 g·mol⁻¹.
- Purity: 99.85% (as supplied).
- Form: solid, off-white to light yellow.
- Solubility: 125 mg/mL in DMSO; ultrasonic assistance recommended.
- Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).
- Available sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg; also offered as 10 mM in DMSO.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.