Learn More
Analytical Sales & Services PFA Film for 24-Position Blocks, Durable Material, 25 Films per Pack
Supplier:  Analytical Sales & Services 24247
Encompass
PFA Film - 24 PSTN Block 25PK is an essential lab consumable designed for 24-position reaction blocks. Made from durable PFA material, it ensures optimal performance in demanding applications. Each pack contains 25 films, allowing for easy and frequent replacements to maintain experimental integrity. Its compatibility with various block assemblies, including photoredox setups, sets it apart, making it indispensable for high-precision laboratory work.
Catalog No. NC0257342