Learn More
Analytical Sales & Services PFA Film For 96-Well Blocks, 25 Films Per Pack, Suitable For Parallel Synthesis
Supplier:  Analytical Sales & Services 96967
Encompass
Replacement PFA films for 96-well blocks streamline laboratory processes by ensuring compatibility with both parallel synthesis and photoredox block assemblies. Supplied in packs of 25, these films offer reliable performance and ease of maintenance, setting them apart in scientific applications. Designed for durability and precision, they enhance experimental outcomes with consistent sealing and protection during various assays.
Catalog No. NC0257346