Learn More
Cayman Chemical PfDHODH-IN-1 10mg
SDP
Supplier:  Cayman Chemical 3772810 MG
Encompass_Preferred
A DHODH inhibitor (IC50s = 190, 47, and 21 nM for the human, mouse, and rat enzymes, respectively); inhibits delayed-type hypersensitivity reactions in mice and rats at 30 and 10 mg/kg, respectively
Catalog No. 50-291-8209