Learn More
Medchemexpress LLC PFDN1 protein, mouse (His) | 2 UG
SDP
Supplier:  Medchemexpress LLC HYP765412UG
Encompass_Preferred
Recombinant mouse prefoldin subunit 1 (PFDN1) with a His tag, expressed in E. coli, intended for biochemical assays and antibody/antigen applications. Supplied in PBS, pH 7.4, the protein has an approximate molecular weight of 16 kDa and low endotoxin (<1 EU/μg). Store at -20°C; after reconstitution, keep at 4°C short term or aliquot and freeze for long-term storage.
  • Recombinant mouse protein with His tag for purification and detection.
  • Expressed in E. coli for high-yield production.
  • Approximate molecular weight 16 kDa for size verification.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in PBS, pH 7.4 for compatibility with common buffers.
  • Stable when stored at -20°C; aliquot and freeze for extended storage.
Catalog No. 50-002-90081