Learn More
ATCC Pfeiffer; Large Cell Lymphoma; Human (Homo sapiens)
Supplier:  ATCC CRL2632
Encompass
Pfeiffer is a B lymphocyte cell line that was isolated in 1992 from a White, adult male patient with diffuse large cell lymphoma, non-Hodgkin's B cell. This cell line was deposited by H Ben-Bassat and can be used in immunology research.
Catalog No. 50-238-2842