Learn More
Selleck Chemical LLC PFI-2 HCl
SDP
Supplier:  Selleck Chemical LLC S729450MG
Encompass_Preferred
PFI-2 is a potent selective and cell-active lysine methyltransferase SETD7 inhibitor with Ki (app) and IC50 of 0 33 nM and 2 nM 1000-fold selectivity over other methyltransferases and other non-epigenetic targets
Catalog No. 50-000-04548