Learn More
Medchemexpress LLC PFI-90 | 53995-62-3 | MFCD27925103 | 99.86% | 214.22 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13934850MG
Encompass_Preferred
PFI-90 is a selective inhibitor of histone demethylase (KDM3B) that inhibits PAX3-FOXO1 action. It induces apoptosis and myogenic differentiation, leading to increased cell death. This compound has potential for antitumor activity.
  • Selective inhibitor of histone demethylase (KDM3B)
  • Inhibits PAX3-FOXO1 action
  • Induces apoptosis and myogenic differentiation
  • Potential for antitumor activity
Catalog No. 50-003-47902