Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 34 (d10A) 200mg
Supplier:  ATCC 511X
Encompass
Purified pneumococcal polysaccharide type 10A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 10A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-3185