Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 56 (d18C) 200mg
Supplier:  ATCC 289X
Encompass
Purified pneumococcal polysaccharide type 18C (US Type 56) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 18C is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 18C can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4792