Learn More
ATCC Pfizer Pneumococcal; polysaccharide powder; Type 71 (38) 200mg
Supplier:  ATCC 544X
Encompass
Purified pneumococcal polysaccharide type 38 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 38 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-0903