Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 57 (d19A) 10mg
Supplier:  ATCC 302X
Encompass
Purified pneumococcal polysaccharide type 19A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 19A is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 19A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-3421