Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 70 (d33F) 200mg
Supplier:  ATCC 535X
Encompass
Purified pneumococcal polysaccharide type 33F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 33F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2312
 Restricted Item