Learn More
ATCC Pfizer Pneumococcal; polysaccharide powder; Type 66 (35B) 200mg
Supplier:  ATCC 541X
Encompass
Purified pneumococcal polysaccharide type 35B is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 35B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2376
 Restricted Item