Learn More
Medchemexpress LLC (E)-1-pyridin-4-yl-3-[7-(trifluoromethyl)quinolin-2-yl]prop-2-en-1-one | 1462249-75-7 | 98.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1220310MM 1ML
Encompass_Preferred
PFK-158 is a small-molecule inhibitor of PFKFB3 used in research on cancer metabolism and related pathways. It is available as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid quantities for custom preparation. The compound has molecular formula C18H11F3N2O, molecular weight 328.29 g/mol, and a reported IC50 of 137 nM.
  • Potent inhibitor of PFKFB3 with reported IC50 of 137 nM.
  • Supplied as 10 mM solution in DMSO (1 mL) and as solid quantities for flexibility.
  • High purity suitable for research (~98.9%).
  • Useful for studies of glucose metabolism, apoptosis, and autophagy in cancer models.
  • Provided for research use only.
Catalog No. 50-002-31175
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.