Learn More
Agilent Technologies PFU DNA POLYMERASE
Supplier:  Agilent Technologies 600140
Encompass
Pfu DNA Polymerases are Ideal for high-fidelity amplification. One of the most thermostable DNA polymerases known. Optimal for blunt-end PCR cloning. Optimum temperature near 75degreeC. 95% active after 1-hour incubation at 98degreeC.
Catalog No. 50-125-921