Learn More
Agilent Technologies PFUULTRA HOTSTART POLYM 500UP
Supplier:  Agilent Technologies 600392
Encompass
DNA polymerase, PfuUltra; Stratagene; Highest fidelity enzyme; Corrects nucleotide misincorporation errors; Available in hotstart mix; Includes ArchaeMaxx; Quantity: 500 U
Catalog No. 50-125-982